Assay Details
| Gene Symbol: LOC105373782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136319.1 | 2 - 3 | 109 | 154 | ||
| GenBank mRNA | BX106730.1 | - | 2 - 3 | 110 | 154 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 804A | ZNF804A | C2orf10 | -225.5kb | Anti-sense | Chr. 2 - 184598366 - 184939487 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927196 | LOC101927196 | 552.9kb | Sense | Chr. 2 - 185719874 - 185740477 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105373782
Genomic Map