Assay Details
| Gene Symbol: LOC105371592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136412.1 | 1 - 2 | 89 | 89 | ||
| GenBank mRNA | BU581060.1 | - | 1 - 2 | 89 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | clustered mitochondria homolog | CLUH |
CLU1, KIAA0664 |
-676.0 | Anti-sense | Chr. 17 - 2689386 - 2711633 on NCBI Build GRCh38 |
| Downstream | coiled-coil domain containing 92B | CCDC92B | 8.0kb | Anti-sense | Chr. 17 - 2720801 - 2748500 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105371592
Genomic Map