Assay Details
| Gene Symbol: LOC105371789 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136415.1 | 1 - 2 | 83 | 67 | ||
| GenBank mRNA | DB338921.1 | - | 2 - 3 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glucose-6-phosphatase catalytic subunit 3 | G6PC3 |
SCN4, UGRP |
-39.6kb | Sense | Chr. 17 - 44070700 - 44076344 on NCBI Build GRCh38 |
| Downstream | chromosome 17 open reading frame 53 | C17orf53 | 21.8kb | Sense | Chr. 17 - 44141902 - 44162477 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105371789
Genomic Map