Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000613453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000613453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000613453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000613453.1 |
Ensembl |