Assay Details
| Gene Symbol: LOC729870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136614.1 | 1 - 2 | 191 | 76 | ||
| GenBank mRNA | BC039551.1 | - | 1 - 2 | 191 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FH2 domain containing 1 | FHDC1 | Anti-sense | Chr. 4 - 152911570 - 152979696 on NCBI Build GRCh38 | ||
| Downstream | tripartite motif containing 2 | TRIM2 |
CMT2R, RNF86 |
215.4kb | Anti-sense | Chr. 4 - 153152281 - 153339322 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC729870
Genomic Map