Assay Details
| Gene Symbol: COL5A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI798815.1 | - | 1 - 2 | 79 | ||
| CR748640.1 | - | 1 - 2 | 130 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | retinoid X receptor alpha | RXRA | NR2B1 | -208.8kb | Anti-sense | Chr. 9 - 134326463 - 134440586 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101448202 | LOC101448202 | 166.6kb | Sense | Chr. 9 - 134819415 - 134872618 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

COL5A1-AS1
Genomic Map