Assay Details
| Gene Symbol: LOC105376432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011519792.2 | XP_011518094.1 | 1 - 2 | 90 | 75 | |
| XM_017017018.1 | XP_016872507.1 | 1 - 2 | 90 | 75 | ||
| XM_017017019.1 | XP_016872508.1 | 1 - 2 | 90 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105376430 | LOC105376430 | -33.8kb | Anti-sense | Chr. 10 - 14972564 - 14988050 on NCBI Build GRCh38 | |
| Downstream | oleoyl-ACP hydrolase | OLAH |
AURA1, SAST, THEDC1 |
8.5kb | Anti-sense | Chr. 10 - 15043896 - 15073852 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011519792.2, XM_017017018.1, XM_017017019.1 transcripts.
Set Membership: |
|

LOC105376432
Genomic Map