Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphodiesterase 4D interacting protein-like | LOC100996724 | Sense | Chr. 1 - 120471449 - 120701470 on NCBI Build GRCh38 | ||
| Downstream | notch homolog 2 N-terminal-like protein | LOC101929796 | 232.1kb | Anti-sense | Chr. 1 - 120723936 - 120804497 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map