Assay Details
| Gene Symbol: FLJ45950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127847.1 | - | 1 - 2 | 145 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tumor protein p53 regulated apoptosis inducing protein 1 | TP53AIP1 | P53AIP1 | -289.5kb | Sense | Chr. 11 - 128934732 - 128944233 on NCBI Build GRCh38 |
| Downstream | BARX homeobox 2 | BARX2 | 92.1kb | Anti-sense | Chr. 11 - 129375237 - 129452279 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK127847.1 transcripts.
Set Membership: |
|

FLJ45950
Genomic Map