Assay Details
| Gene Symbol: SLC16A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103743.1 | 2 - 3 | 1405 | 89 | ||
| NR_103744.1 | 2 - 3 | 639 | 89 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 16 member 1 | SLC16A1 |
HHF7, MCT, MCT1, MCT1D |
-4.1kb | Anti-sense | Chr. 1 - 112911847 - 112956353 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100996251 | LOC100996251 | 47.6kb | Anti-sense | Chr. 1 - 113011687 - 113073102 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000420168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000420168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000420168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000420168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000420168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | NR_103744.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | NR_103744.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | NR_103744.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | NR_103744.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | NR_103744.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | NR_103744.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009463.hg.1 | NR_103744.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000416193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000420168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000420168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000420168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000420168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000420168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000428411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | ENST00000435800.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | NR_103743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | NR_103744.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | NR_103744.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | NR_103744.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | NR_103744.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | NR_103744.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | NR_103744.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009463.hg.1 | NR_103744.1 | Refseq |

SLC16A1-AS1
Genomic Map