Assay Details
| Gene Symbol: FMR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024503.3 | 1 - 2 | 180 | 95 | ||
| GenBank mRNA | EU251068.1 | - | 1 - 2 | 180 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 514a-3 | MIR514A3 |
MIR514-3, MIRN514-3, hsa-mir-514a-3 |
-624.7kb | Sense | Chr. X - 147284641 - 147284728 on NCBI Build GRCh38 |
| Downstream | fragile X mental retardation 1 | FMR1 |
FMRP, FRAXA, POF, POF1 |
134.0 | Anti-sense | Chr. X - 147911951 - 147951127 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

FMR1-AS1
Genomic Map