Assay Details
| Gene Symbol: MIR588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030316.1 | 1 | 43 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | centromere protein W | CENPW |
C6orf173, CENP-W, CUG2 |
-1.3kb | Sense | Chr. 6 - 126339696 - 126483320 on NCBI Build GRCh38 |
| Downstream | R-spondin 3 | RSPO3 |
CRISTIN1, PWTSR, THSD2 |
634.0kb | Sense | Chr. 6 - 127118671 - 127199481 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

MIR588
Genomic Map