Assay Details
| Gene Symbol: LOC100289673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033175.1 | 2 - 3 | 197 | 109 | ||
| GenBank mRNA | AI125295.1 | - | 1 - 2 | 109 | ||
| BX093587.1 | - | 2 - 3 | 196 | 109 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mannosidase alpha class 2A member 1 | MAN2A1 |
AMan II, GOLIM7, MANA2, MANII |
-14.2kb | Sense | Chr. 5 - 109689366 - 109869625 on NCBI Build GRCh38 |
| Downstream | transmembrane protein 232 | TMEM232 | 501.8kb | Anti-sense | Chr. 5 - 110387255 - 110738936 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC100289673
Genomic Map