Assay Details
| Gene Symbol: MIR3137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036089.1 | 1 | 35 | 55 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | XXYLT1 antisense RNA 1 | XXYLT1-AS1 | -38.4kb | Anti-sense | Chr. 3 - 195094588 - 195096057 on NCBI Build GRCh38 | |
| Downstream | XXYLT1 antisense RNA 2 | XXYLT1-AS2 | 13.3kb | Anti-sense | Chr. 3 - 195147871 - 195152856 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

MIR3137
Genomic Map