Assay Details
| Gene Symbol: MIR6165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106751.1 | 1 | 37 | 62 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927207 | LOC101927207 | -49.1kb | Anti-sense | Chr. 17 - 49457861 - 49461749 on NCBI Build GRCh38 | |
| Downstream | neurexophilin 3 | NXPH3 | NPH3 | 65.0kb | Sense | Chr. 17 - 49575936 - 49583810 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

MIR6165
Genomic Map