Assay Details
| Gene Symbol: MIR1273H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106996.1 | 1 | 59 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calcineurin like EF-hand protein 2 | CHP2 | -444.2kb | Sense | Chr. 16 - 23754627 - 23758951 on NCBI Build GRCh38 | |
| Downstream | calcium voltage-gated channel auxiliary subunit gamma 3 | CACNG3 | 52.3kb | Sense | Chr. 16 - 24255553 - 24362416 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

MIR1273H
Genomic Map