| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000471299.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000471299.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000471299.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000471299.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000479592.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000479592.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000479592.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000479592.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000492208.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000492208.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
ENST00000492208.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000471299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000471299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000471299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000471299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000479592.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000479592.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000479592.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000479592.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000484550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000492208.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000492208.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
ENST00000492208.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006442.hg.1 |
NR_108064.1 |
Refseq |