Assay Details
| Gene Symbol: LOC101929549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110112.1 | 3 - 4 | 488 | 163 | ||
| GenBank mRNA | BC043558.1 | - | 3 - 4 | 488 | 163 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NADH:ubiquinone oxidoreductase subunit A9 | NDUFA9 |
CC6, CI-39k, CI39k, NDUFS2L, SDR22E1 |
-12.9kb | Anti-sense | Chr. 12 - 4649098 - 4687554 on NCBI Build GRCh38 |
| Downstream | polypeptide N-acetylgalactosaminyltransferase 8 | GALNT8 | GALNAC-T8 | 17.1kb | Anti-sense | Chr. 12 - 4720586 - 4772726 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101929549
Genomic Map