Assay Details
| Gene Symbol: LINC01461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125761.1 | 1 - 2 | 281 | 71 | ||
| GenBank mRNA | DA091614.1 | - | 1 - 2 | 282 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chitobiase | CTBS | CTB | -44.3kb | Sense | Chr. 1 - 84553121 - 84574480 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1555 | LINC01555 | C1orf180 | 7.2kb | Sense | Chr. 1 - 84628230 - 84635020 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01461
Genomic Map