Assay Details
| Gene Symbol: LOC102723604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126331.1 | 2 - 3 | 168 | 124 | ||
| GenBank mRNA | AI693930.1 | - | 1 - 2 | 124 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927598 | LOC101927598 | -91.1kb | Anti-sense | Chr. 14 - 51765816 - 51825019 on NCBI Build GRCh38 | |
| Downstream | chromosome 14 open reading frame 166 | C14orf166 |
CGI-99, CGI99, CLE, CLE7, LCRP369, RLLM1, hCLE1 |
72.0kb | Anti-sense | Chr. 14 - 51989510 - 52004702 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC102723604
Genomic Map