Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000525941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000525941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000525941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000525941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000525941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000525941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000526588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000526588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000526588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000526588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000526588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000530083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000530083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000530083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000530083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000530083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000533938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000533938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000623692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000623846.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
ENST00000623846.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
NR_126342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
NR_126342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
NR_126342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
NR_126342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
NR_126342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
NR_126342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
NR_126342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
NR_126343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
NR_126343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
NR_126343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
NR_126343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
NR_126343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006488.hg.1 |
NR_126343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000525941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000525941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000525941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000525941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000525941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000525941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000526588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000526588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000526588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000526588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000526588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000530083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000530083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000530083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000530083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000530083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000533938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000533938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000623692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000623846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
ENST00000623846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
NR_126342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
NR_126342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
NR_126342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
NR_126342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
NR_126342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
NR_126342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
NR_126342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
NR_126343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
NR_126343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
NR_126343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
NR_126343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
NR_126343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006488.hg.1 |
NR_126343.1 |
Refseq |