Assay Details
| Gene Symbol: LINC01389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126355.1 | 3 - 4 | 406 | 97 | ||
| GenBank mRNA | AI393217.1 | - | 2 - 3 | 97 | ||
| BX102452.1 | - | 1 - 2 | 171 | 97 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cytidine/uridine monophosphate kinase 1 | CMPK1 |
CK, CMK, CMPK, UMK, UMP-CMPK, UMPK |
-7.4kb | Anti-sense | Chr. 1 - 47333797 - 47378839 on NCBI Build GRCh38 |
| Downstream | forkhead box E3 | FOXE3 |
FKHL12, FREAC8 |
7.6kb | Anti-sense | Chr. 1 - 47416072 - 47418052 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01389
Genomic Map