Assay Details
| Gene Symbol: SALRNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126482.1 | 1 | 1918 | 105 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | senescence associated long non-coding RNA 3 | SALRNA3 |
SAL-RNA3, XLOC_025918 |
-18.7kb | Sense | Chr. 15 - 70615547 - 70616567 on NCBI Build GRCh38 |
| Downstream | uveal autoantigen with coiled-coil domains and ankyrin repeats | UACA | NUCLING | 17.2kb | Anti-sense | Chr. 15 - 70654554 - 70778823 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SALRNA2
Genomic Map