Assay Details
Gene Symbol: LOC606724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002454.2 | 1 | 516 | 107 | ||
GenBank mRNA | U35342.1 | - | 1 | 516 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
