Assay Details
| Gene Symbol: LOC105374698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134265.1 | 2 | 225 | 120 | ||
| GenBank mRNA | BX112617.1 | - | 1 | 114 | 120 | |
| CR740698.1 | - | 2 | 225 | 120 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1021 | LINC01021 | -790.0kb | Anti-sense | Chr. 5 - 27472292 - 27496401 on NCBI Build GRCh38 | |
| Downstream | lymphocyte-specific protein 1 pseudogene 3 | LSP1P3 | 640.0kb | Anti-sense | Chr. 5 - 28926870 - 28927313 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105374698
Genomic Map