Assay Details
| Gene Symbol: LOC105376398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134492.1 | 2 | 224 | 78 | ||
| GenBank mRNA | BX100511.1 | - | 2 | 224 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 708 | LINC00708 | GS1-756B1.2 | -304.4kb | Sense | Chr. 10 - 8259331 - 8268305 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105755953 | LOC105755953 | 324.6kb | Anti-sense | Chr. 10 - 8897989 - 8914596 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105376398
Genomic Map