Assay Details
| Gene Symbol: SNORD130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132973.1 | 1 | 51 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | membrane palmitoylated protein 7 | MPP7 | Sense | Chr. 10 - 28050993 - 28303066 on NCBI Build GRCh38 | ||
| Downstream | microRNA 8086 | MIR8086 | hsa-mir-8086 | 215.9kb | Sense | Chr. 10 - 28289258 - 28289350 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SNORD130
Genomic Map