Assay Details
| Gene Symbol: SNORA101A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135619.1 | 1 | 66 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929353 | LOC101929353 | -179.6kb | Anti-sense | Chr. 4 - 100190033 - 100215505 on NCBI Build GRCh38 | |
| Downstream | endomucin | EMCN |
EMCN2, MUC14 |
94.0 | Sense | Chr. 4 - 100395341 - 100518093 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SNORA101A
Genomic Map