Assay Details
| Gene Symbol: LOC105747689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130181.1 | - | 1888 | 58 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nucleic acid binding protein 1 | NABP1 |
OBFC2A, SOSS-B2, SSB2 |
-7.7kb | Anti-sense | Chr. 2 - 191678072 - 191688522 on NCBI Build GRCh38 |
| Downstream | hCG2045843 | LOC729254 | 96.2kb | Anti-sense | Chr. 2 - 191792840 - 191820250 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105747689
Genomic Map