Assay Details
| Gene Symbol: LINC01123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046110.1 | 2 - 3 | 1448 | 80 | ||
| GenBank mRNA | AK056084.1 | - | 2 - 3 | 1365 | 80 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200013869.hg.1 | ENST00000436665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013869.hg.1 | ENST00000436665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013869.hg.1 | ENST00000436665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013869.hg.1 | ENST00000436665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013869.hg.1 | ENST00000436665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013869.hg.1 | ENST00000436665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013869.hg.1 | ENST00000448359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013869.hg.1 | ENST00000448359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013869.hg.1 | ENST00000448359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013869.hg.1 | ENST00000448359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013869.hg.1 | ENST00000448359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013869.hg.1 | NR_027244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013869.hg.1 | NR_027244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013869.hg.1 | NR_027244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013869.hg.1 | NR_027244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013869.hg.1 | NR_027244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013869.hg.1 | NR_027244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000336905.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000336905.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000336905.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000336905.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000336905.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000336905.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000336905.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000336905.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008830.hg.1 | NR_046112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000336905.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000336905.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000336905.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000336905.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000336905.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000336905.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000336905.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000336905.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | ENST00000419296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008830.hg.1 | NR_046112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013869.hg.1 | ENST00000436665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013869.hg.1 | ENST00000436665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013869.hg.1 | ENST00000436665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013869.hg.1 | ENST00000436665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013869.hg.1 | ENST00000436665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013869.hg.1 | ENST00000436665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013869.hg.1 | ENST00000448359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013869.hg.1 | ENST00000448359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013869.hg.1 | ENST00000448359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013869.hg.1 | ENST00000448359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013869.hg.1 | ENST00000448359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013869.hg.1 | NR_027244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013869.hg.1 | NR_027244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013869.hg.1 | NR_027244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013869.hg.1 | NR_027244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013869.hg.1 | NR_027244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013869.hg.1 | NR_027244.1 | Refseq |

LINC01123
Genomic Map