Assay Details
| Gene Symbol: C22orf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017028796.1 | XP_016884285.1 | 6 - 7 | 9845 | 79 | |
| GenBank mRNA | AK128192.1 | - | 1 - 2 | 136 | 79 | |
More Information
Set Membership: |
|

C22orf34
Genomic Map