Assay Details
| Gene Symbol: SIAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011523279.1 | XP_011521581.1 | 4 - 5 | 1416 | 87 | |
| GenBank mRNA | BC044920.1 | - | 4 - 5 | 1379 | 5295925 | 87 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC044920.1, XM_011523279.1 transcripts.
Set Membership: |
|

SIAH1
Genomic Map