| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
ENST00000565039.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
ENST00000565039.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
ENST00000565039.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
ENST00000565039.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
ENST00000565039.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
ENST00000336257.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
ENST00000565039.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
ENST00000565039.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
ENST00000565039.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
ENST00000565039.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
ENST00000565039.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011567.hg.1 |
NM_024533.4 |
Refseq |