Assay Details
| Gene Symbol: SNAI3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024402.2 | 3 - 4 | 521 | 63 | ||
| GenBank mRNA | BX640980.1 | - | 3 - 4 | 541 | 63 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000565633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000565633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000565633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000565633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000565633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000565633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000567997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000567997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000569786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000569786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000569786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000569786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000569786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000569786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000596908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | ENST00000596908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | NR_024399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | NR_024399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | NR_024399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000563475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000565633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000565633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000565633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000565633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000565633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000565633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000567997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000567997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000568633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000569786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000569786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000569786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000569786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000569786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000569786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000596908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | ENST00000596908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | NR_024399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | NR_024399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | NR_024399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | NR_024402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008834.hg.1 | NR_024402.2 | Refseq |

SNAI3-AS1
Genomic Map