Assay Details
| Gene Symbol: RN7SKP23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK126661.1 | - | 1 | 1391 | 101 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600007220.hg.1 | ENST00000625159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007220.hg.1 | ENST00000625159.1 | Ensembl |

RN7SKP23
Genomic Map