Assay Details
| Gene Symbol: CSNK2A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090752.1 | - | 1 | 272 | 101 | |
| AK125922.1 | - | 2 | 3068 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSNK2A2| Gene Symbol: CSNK2A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090752.1 | - | 1 | 272 | 101 | |
| AK125922.1 | - | 2 | 3068 | 101 | ||
Set Membership: |
|