Assay Details
| Gene Symbol: MAPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109891.1 | NP_001103361.1 | 8 | 1519 | 78 | |
| NM_002746.2 | NP_002737.2 | 9 | 1651 | 78 | ||
| GenBank mRNA | AK091009.1 | - | 8 | 2343 | 78 | |
| BC000205.1 | - | 4 | 771 | 78 | ||
| BC013992.1 | - | 9 | 1562 | 3634492 | 78 | |
| BX537897.1 | - | 8 | 2342 | 78 | ||
| M84490.1 | - | 9 | 1510 | 78 | ||
| X60188.1 | - | 10 | 1626 | 78 | ||
More Information
Set Membership: |
|

MAPK3
Genomic Map