Assay Details
| Gene Symbol: SLC9A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021876.1 | - | 1 | 1081 | 87 | |
| AK091748.1 | - | 1 | 1947 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC9A5| Gene Symbol: SLC9A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021876.1 | - | 1 | 1081 | 87 | |
| AK091748.1 | - | 1 | 1947 | 87 | ||
Set Membership: |
|