Assay Details
Gene Symbol: LINC00922 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027755.1 | 9 | 924 | 135 | ||
GenBank mRNA | BC037902.1 | - | 9 | 930 | 135 | |
BC104446.1 | - | 10 | 932 | 135 | ||
BC104447.1 | - | 6 | 533 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000568492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000568492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000568492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000568492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000568492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000568492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000564041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000568492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000568492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000568492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000568492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000568492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000568492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | ENST00000569736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | NR_027755.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010567.hg.1 | NR_027755.1 | Refseq |
