Assay Details
| Gene Symbol: PIGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017025352.1 | XP_016880841.1 | 8 - 9 | 1931 | 156 | |
| XM_017025354.1 | XP_016880843.1 | 7 - 8 | 1899 | 156 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700012199.hg.1 | ENST00000414755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012199.hg.1 | ENST00000414755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012199.hg.1 | ENST00000414755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012199.hg.1 | ENST00000431149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012199.hg.1 | ENST00000431149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012199.hg.1 | ENST00000431149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012199.hg.1 | ENST00000431149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012199.hg.1 | ENST00000431149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012199.hg.1 | ENST00000431149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000631687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000631687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000631687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000631687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012199.hg.1 | ENST00000414755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012199.hg.1 | ENST00000414755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012199.hg.1 | ENST00000414755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012199.hg.1 | ENST00000431149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012199.hg.1 | ENST00000431149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012199.hg.1 | ENST00000431149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012199.hg.1 | ENST00000431149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012199.hg.1 | ENST00000431149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012199.hg.1 | ENST00000431149.5 | Ensembl |

PIGL
Genomic Map