Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010668.hg.1 |
ENST00000393988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010668.hg.1 |
ENST00000393988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010668.hg.1 |
ENST00000393988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010668.hg.1 |
ENST00000393988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010668.hg.1 |
ENST00000393988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010668.hg.1 |
ENST00000393988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010668.hg.1 |
ENST00000393988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007834.hg.1 |
ENST00000430006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007834.hg.1 |
ENST00000430006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007834.hg.1 |
ENST00000430006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007834.hg.1 |
ENST00000430006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007834.hg.1 |
ENST00000430006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007834.hg.1 |
ENST00000430006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007834.hg.1 |
ENST00000432258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007834.hg.1 |
ENST00000432258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007834.hg.1 |
ENST00000432258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007834.hg.1 |
ENST00000432258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007834.hg.1 |
NR_040111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007834.hg.1 |
NR_040111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007834.hg.1 |
NR_040111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010668.hg.1 |
ENST00000393988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010668.hg.1 |
ENST00000393988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010668.hg.1 |
ENST00000393988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010668.hg.1 |
ENST00000393988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010668.hg.1 |
ENST00000393988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010668.hg.1 |
ENST00000393988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010668.hg.1 |
ENST00000393988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007834.hg.1 |
ENST00000430006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007834.hg.1 |
ENST00000430006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007834.hg.1 |
ENST00000430006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007834.hg.1 |
ENST00000430006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007834.hg.1 |
ENST00000430006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007834.hg.1 |
ENST00000430006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007834.hg.1 |
ENST00000432258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007834.hg.1 |
ENST00000432258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007834.hg.1 |
ENST00000432258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007834.hg.1 |
ENST00000432258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007834.hg.1 |
NR_040111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007834.hg.1 |
NR_040111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007834.hg.1 |
NR_040111.1 |
Refseq |