Assay Details
| Gene Symbol: RNF135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184992.1 | NP_001171921.1 | 3 - 4 | 814 | 102 | |
| GenBank mRNA | BC082262.1 | - | 3 - 4 | 692 | 102 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
protein ubiquitination
negative regulation of type I interferon production
positive regulation of interferon-beta production
innate immune response
regulation of innate immune response
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ribonucleoprotein complex binding
protein ubiquitination
negative regulation of type I interferon production
positive regulation of interferon-beta production
innate immune response
regulation of innate immune response
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ribonucleoprotein complex binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000434242.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000434242.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000434242.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000434242.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000443677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000443677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000443677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000443677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000443677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000443677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000443677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000443677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000580444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000580444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000580444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000582674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | ENST00000582674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000324689.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000328381.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000434242.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000434242.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000434242.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000434242.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000443677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000443677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000443677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000443677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000443677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000443677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000443677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000443677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000535306.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000580444.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000580444.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000580444.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000582674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | ENST00000582674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_001184992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_032322.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_197939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007474.hg.1 | NM_197939.1 | Refseq |

RNF135
Genomic Map