Assay Details
| Gene Symbol: RNF135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184992.1 | NP_001171921.1 | 4 - 5 | 882 | 84 | |
| GenBank mRNA | BC082262.1 | - | 4 - 5 | 760 | 84 | |
More Information
Set Membership: |
|

RNF135
Genomic Map