Assay Details
| Gene Symbol: DUSP28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033575.1 | NP_001028747.1 | 2 - 3 | 1219 | 99 | |
| GenBank mRNA | BQ777661.1 | - | 1 - 2 | 99 | ||
More Information
Set Membership: |
|

DUSP28
Genomic Map