Assay Details
| Gene Symbol: LINC00211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DB092735.1 | - | 2 - 3 | 326 | 125 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000418746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000418746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000418746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000418746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000418746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000418746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110012.1 | Refseq |

LINC00211
Genomic Map