Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012893.hg.1 |
ENST00000439433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012893.hg.1 |
ENST00000439433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012893.hg.1 |
ENST00000439433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012893.hg.1 |
ENST00000454595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012893.hg.1 |
ENST00000454595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012893.hg.1 |
NR_046625.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012893.hg.1 |
NR_046625.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012893.hg.1 |
NR_046625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012893.hg.1 |
ENST00000439433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012893.hg.1 |
ENST00000439433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012893.hg.1 |
ENST00000439433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012893.hg.1 |
ENST00000454595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012893.hg.1 |
ENST00000454595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012893.hg.1 |
NR_046625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012893.hg.1 |
NR_046625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012893.hg.1 |
NR_046625.1 |
Refseq |