Assay Details
| Gene Symbol: SDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006946.1 | NP_001006947.1 | 3 - 4 | 542 | 98 | |
| NM_002997.4 | NP_002988.3 | 2 - 3 | 450 | 98 | ||
| XM_005262620.4 | XP_005262677.1 | 2 - 3 | 293 | 98 | ||
| XM_005262621.3 | XP_005262678.3 | 2 - 3 | 173 | 98 | ||
| XM_005262622.2 | XP_005262679.1 | 2 - 3 | 112 | 98 | ||
| GenBank mRNA | AJ551176.1 | - | 2 - 3 | 399 | 98 | |
| AK222739.1 | - | 2 - 3 | 381 | 98 | ||
| AK299385.1 | - | 2 - 3 | 351 | 98 | ||
| AK307313.1 | - | 2 - 3 | 271 | 98 | ||
| AK313974.1 | - | 2 - 3 | 378 | 98 | ||
| BC008765.2 | - | 2 - 3 | 394 | 3347793 | 98 | |
| BM910157.1 | - | 452 | 98 | |||
| J05392.1 | - | 2 - 3 | 356 | 98 | ||
| X60306.1 | - | 2 - 3 | 151 | 98 | ||
More Information
Set Membership: |
|

SDC1
Genomic Map