Assay Details
| Gene Symbol: PUM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011532719.1 | XP_011531021.1 | 3 - 4 | 427 | 108 | |
| XM_017003704.1 | XP_016859193.1 | 3 - 4 | 449 | 108 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011532719.1, XM_017003704.1 transcripts.
Set Membership: |
|

PUM2
Genomic Map