Assay Details
| Gene Symbol: RTN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005264434.3 | XP_005264491.1 | 1 - 2 | 98 | 177 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005264434.3 transcripts.
Set Membership: |
|

RTN4
Genomic Map