Assay Details
| Gene Symbol: RTN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321904.1 | NP_001308833.1 | 2 - 3 | 546 | 92 | |
| XM_005264434.3 | XP_005264491.1 | 2 - 3 | 245 | 92 | ||
| XM_017004518.1 | XP_016860007.1 | 2 - 3 | 284 | 92 | ||
| XM_017004519.1 | XP_016860008.1 | 2 - 3 | 401 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001321904.1, XM_005264434.3, XM_017004518.1, XM_017004519.1 transcripts.
Set Membership: |
|

RTN4
Genomic Map